Anti-PFK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-3331R-FITC
10367-504EA 776.17 USD
10367-504
Anti-PFK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
6 Phosphofructo 2 Kinase plays a role in the synthesis and degradation of fructose 2,6-bisphosphate. Fructose-2,6-bisphosphate acts as an allosteric regulator of 6-phosphofructo-1-kinase (PFK1), which catalyses a key step in the glycoytic pathway. Glycolytic flux is key to tumour growth, and small molecule inhibition of 6 Phosphofructo 2 Kinase suppresses both glycolytic flux and tumour growth. Several lines of evidence suggest that 6 Phosphofructo 2 Kinase is also involved in obesity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PFK2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR