6 Phosphofructo 2 Kinase plays a role in the synthesis and degradation of fructose 2,6-bisphosphate. Fructose-2,6-bisphosphate acts as an allosteric regulator of 6-phosphofructo-1-kinase (PFK1), which catalyses a key step in the glycoytic pathway. Glycolytic flux is key to tumour growth, and small molecule inhibition of 6 Phosphofructo 2 Kinase suppresses both glycolytic flux and tumour growth. Several lines of evidence suggest that 6 Phosphofructo 2 Kinase is also involved in obesity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PFK2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat