Anti-NUMA Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3312R-CY5.5
10369-902EA 776.19 USD
10369-902
Anti-NUMA Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
NuMA (Nuclear Mitotic Apparatus Protein) is an intranuclear protein and present in nucleus during interphase. At the onset of mitosis, it redistributes from the nucleus to two centrosomal structures that later will become part of the mitotic spindle pole. After anaphase, the protein redistributes from the spindle polar region into reforming nucleus. NuMA is an essential protein during mitosis for the terminal phases of chromosome separation and/or nuclear reassembly. Recently a study shows that NuMA is cleaved to a 180 to 200kDa during apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NUMA
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR