Anti-DUSP1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-3278R-CY7
10366-704EA 776.19 USD
10366-704
Anti-DUSP1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The expression of DUSP1 gene is induced in human skin fibroblasts by oxidative/heat stress and growth factors. It specifies a protein with structural features similar to members of the non-receptor-type protein-tyrosine phosphatase family, and which has significant amino-acid sequence similarity to a Tyr/Ser-protein phosphatase encoded by the late gene H1 of vaccinia virus. The bacterially expressed and purified DUSP1 protein has intrinsic phosphatase activity, and specifically inactivates mitogen-activated protein (MAP) kinase in vitro by the concomitant dephosphorylation of both its phosphothreonine and phosphotyrosine residues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DUSP1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR