Anti-MARCKS Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-3263R-FITC
10366-442EA 775.95 USD
10366-442
Anti-MARCKS Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
MARCKS, (Myristoylated Alanine-Rich C Kinase Substrate), is a member of a family of calmodulin binding proteins and is a major substrate for phosphorylation by protein kinase C (PKC). The phosphorylation of Ser152/156 can be used as a measure of PKC activation. Phosphorylation of Ser152/156 modulates the binding of MARCKS to calmodulin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MARCKS
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR