Anti-JAK1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3238R-A555
10371-916EA 776.19 USD
10371-916
Anti-JAK1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Tyrosine kinase of the non-receptor type, involved in the IFN-alpha/beta/gamma signal pathway. Kinase partner for the interleukin (IL)-2 receptor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: JAK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR