Anti-CBX3 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3221R-CY5
10355-884EA 811.12 USD
10355-884
Anti-CBX3 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
HP1 gamma appears to be involved in transcriptional silencing in heterochromatin-like complexes. It recognizes and binds histone H3 tails methylated at 'Lys-9', leading to epigenetic repression. HP1 gamma may contribute to the association of the heterochromatin with the inner nuclear membrane through its interaction with lamin B receptor (LBR) and involved in the formation of functional kinetochore through interaction with MIS12 complex proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CBX3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR