|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | NFKBIB |
Antigen Name | Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells Inhibitor, Beta |
Reactivity | Rat, Human, Mouse |
Conjugation | FITC (Fluorescein Isothiocyanate) |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | IkBb, NF-kappa-BIB, IKB-beta, ikB-B, I(Kappa)B(beta), IKappaBbeta, IkB, I-kappa-B-beta |
Gene ID | 4793 |
Modification | phospho-Ser23 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |