Anti-IKB beta Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3191R-A488
10353-892EA 811.12 USD
10353-892
Anti-IKB beta Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
The protein encoded by this gene belongs to the NF-kappa-B inhibitor family, which inhibit NF-kappa-B by complexing with, and trapping it in the cytoplasm. Phosphorylation of serine residues on these proteins by kinases marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B, which translocates to the nucleus to function as a transcription factor. Alternatively spliced transcript variants have been found for this gene.[provided by RefSeq, Jul 2011].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IKB beta
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR