Anti-HIST3H3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-3189R
10353-856EA 649.66 USD
10353-856
Anti-HIST3H3 Rabbit Polyclonal Antibody
Antibodies
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fibre is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. Covalent modifications of the canonical core histones, including acetylation, phosphorylation, methylation, and monoubiquitination are used to mark nucleosomes to create chromatin domains with a range of functions. The information encoded by histone modifications can contribute to the formation and/or maintenance of transcriptionally active and inactive chromatin in response to various signalling pathways.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: HIST3H3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR