|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | H2AFX |
Antigen Name | H2A histone Family, member X |
Reactivity | Rat, Human, Mouse |
Conjugation | Cy3® |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | H2A histone family member X, H2AX, H2A.x, histone H2A, gamma-H2AX, H2AFX anitbody |
Gene ID | 3014 |
Modification | phospho-Ser140 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 100-142/143 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |