Anti-GAB2 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-3183R-CY5
10353-726EA 811.12 USD
10353-726
Anti-GAB2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Gab2 (Grb2-associated binder-2) is an adaptor protein involved in numerous intracellular signaling pathways via its interaction with receptor tyrosine kinases (RTKs), growth factors and cytokines. Gab2 is a substrate of various RTKs, including EGFR, insulin receptor, cytokine receptors as well as B and T cell antigen receptors; Gab2 acts as a docking protein for several SH2-containing proteins. Upon tyrosine phosphorylation, Gab2 interacts with SHP2 tyrosine phosphatase and GRB2 adaptor protein PI3-kinase, CrkL and SHC.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GAB2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR