|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | GAB2 |
Antigen Name | GRB2-associated binding protein 2 |
Reactivity | Human, Rat, Mouse |
Conjugation | Cy3® |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | growth factor receptor bound protein 2-associated protein 2, GRB2-associated-binding protein 2, Grb2-associated binder 2, pp100 |
Gene ID | 9846 |
Modification | phospho-Tyr643 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |