Anti-HSP90AA1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3181R-CY3
10355-762EA 775.95 USD
10355-762
Anti-HSP90AA1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Molecular chaperone that promotes the maturation, structural maintenance and proper regulation of specific target proteins involved for instance in cell cycle control and signal transduction. Undergoes a functional cycle that is linked to its ATPase activity. This cycle probably induces conformational changes in the client proteins, thereby causing their activation. Interacts dynamically with various co-chaperones that modulate its substrate recognition, ATPase cycle and chaperone function. Binds bacterial lipopolysaccharide (LPS) et mediates LPS-induced inflammatory response, including TNF secretion by monocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSP90AA1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR