Anti-NR3C1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-3168R-CY7
10353-506EA 811.12 USD
10353-506
Anti-NR3C1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Steroid receptors are ligand-dependent, intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. Glucocorticoids are a family of steroids necessary for the regulation of energy metabolism and the immune and inflammatory responses. These compounds exert their effect through their interaction with the glucocoticoid receptor (GR) and that complex's subsequent association with DNA. All normal mammalian tissues examined to date have been shown to contain glucocorticoid receptor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NR3C1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR