|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PTK2 |
Antigen Name | Protein-tyrosine Kinase 2 |
Reactivity | Human, Mouse, Rat |
Conjugation | Alexa Fluor® 647 |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | FAK-related non-kinase polypeptide, Focal Adhesion Kinase, protein-tyrosine kinase 2, focal adhesion kinase 1, FADK 1 |
Gene ID | 5747 |
Modification | phospho-Tyr861 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 840-890/1052 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |