|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | EIF2AK4 |
Antigen Name | eukaryotic translation initiation factor 2 alpha kinase 4 |
Reactivity | Human, Rat, Mouse |
Conjugation | Alexa Fluor® 555 |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | PVOD2, GCN2 |
Gene ID | 440275 |
Modification | phospho-Thr667 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 640-690/1649 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |