Anti-EIF2AK4 Rabbit Polyclonal Antibody (Biotin)

BS-3155R-BIO
10351-922EA 575.55 USD
10351-922
Anti-EIF2AK4 Rabbit Polyclonal Antibody (Biotin)
GCN2 belongs to a family of kinases that phosphorylate the alpha subunit of eukaryotic translation initiation factor 2 to downregulate protein synthesis in response to varied cellular stresses.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: EIF2AK4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR