Anti-EIF2AK4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3155R-A488
10351-916EA 776.17 USD
10351-916
Anti-EIF2AK4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
GCN2 belongs to a family of kinases that phosphorylate the alpha subunit of eukaryotic translation initiation factor 2 to downregulate protein synthesis in response to varied cellular stresses.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EIF2AK4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR