Anti-GSK3B Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-3148R-CY7
10355-648EA 811.12 USD
10355-648
Anti-GSK3B Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Glycogen synthase kinase 3 (GSK3) is a proline directed serine threonine kinase that was initially identified as a phosphorylating and inactivating glycogen synthase. GSK3 has been implicated in fundamental cell processes such as cell fate determination, metabolism, transcriptional control and oncogenesis. Two isoforms, alpha (GSK3A; OMIM 606784) and beta, show a high degree of amino acid homology within their catalytic domains. GSK3B is involved in energy metabolism, neuronal cell development and body pattern formation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GSK-3 Beta
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR