Anti-FOXO3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3140R-A555
10351-742EA 811.1 USD
10351-742
Anti-FOXO3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Transcriptional activator which triggers apoptosis in the absence of survival factors, including neuronal cell death upon oxidative stress. Recognizes and binds to the DNA sequence 5'-[AG]TAAA[TC]A-3'. Participates in post-transcriptional regulation of MYC: following phosphorylation by MAPKAPK5, promotes induction of miR-34b and miR-34c expression, 2 post-transcriptional regulators of MYC that bind to the 3'UTR of MYC transcript and prevent its translation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FoxO3a
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR