|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | BMX |
Antigen Name | BMX non-receptor tyrosine kinase |
Reactivity | Rat, Mouse, Human |
Conjugation | Cy3® |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | Cytoplasmic tyrosine-protein kinase BMX, Bone marrow kinase BMX, Epithelial and endothelial tyrosine kinase, NTK38, ETK |
Gene ID | 660 |
Modification | phospho-Tyr40 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |