Anti-CCNB1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3123R-CY3
10355-602EA 811.12 USD
10355-602
Anti-CCNB1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
The protein encoded by this gene is a regulatory protein involved in mitosis. The gene product complexes with p34(cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predominantly during G2/M phase. The different transcripts result from the use of alternate transcription initiation sites. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCNB1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR