Anti-CRK Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3121R-CY5.5
10351-440EA 811.12 USD
10351-440
Anti-CRK Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
The Crk-I and Crk-II forms differ in their biological activities. Crk-II has less transforming activity than Crk-I. Crk-II mediates attachment-induced MAPK8 activation, membrane ruffling and cell motility in a Rac-dependent manner. Involved in phagocytosis of apoptotic cells and cell motility via its interaction with DOCK1 and DOCK4. May regulate the EFNA5-EPHA3 signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRK
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR