Anti-CRK Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3121R-A555
10351-430EA 811.12 USD
10351-430
Anti-CRK Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
The Crk-I and Crk-II forms differ in their biological activities. Crk-II has less transforming activity than Crk-I. Crk-II mediates attachment-induced MAPK8 activation, membrane ruffling and cell motility in a Rac-dependent manner. Involved in phagocytosis of apoptotic cells and cell motility via its interaction with DOCK1 and DOCK4. May regulate the EFNA5-EPHA3 signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRK
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR