Anti-PPP1R1B Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3118R-CY5.5
10353-064EA 811.12 USD
10353-064
Anti-PPP1R1B Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Inhibitor of protein-phosphatase 1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PPP1R1B
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR