Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DAB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-3116R-HRP
Disabled 1 (Dab1) is an 80 kDa protein that is encoded by the Disabled-1 gene locus which is mutated in scrambler and yotari mutant mice. Phenotypically, the mutation of this gene produces motor defects and ataxia, disruption of neuronal migration, and severe cerebellar hypoplasia. Dab1 is an intracellular adapter protein that functions in downstream signaling events initiated by the secreted protein reelin. Dab1 contains a phosphotyrosine binding (PTB) domain in the amino terminus. Tyrosine phosphorylation of Dab1 is increased by reelin binding to the Very Low Density Lipoprotein Receptor (VLDLR) and Apolipoprotein E Receptor 2 (ApoER2) through stimulation of Src family kinases. Src family kinase and c-Abl activities are themselves then stimulated by binding to tyrosine phosphorylated Dab1. Dab1 also mediates activation of Akt (PKB) by reelin resulting in inhibition of glycogen synthase kinase 3 beta (GSK-3 beta) and decreased phosphorylation of the microtubule-associated protein, Tau. Dab1 serine 491 is phosphorylated in a Cdk5-dependent manner and regulates, likely indirectly, Reelin-induced signaling during neural cortex development.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: DAB1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-3116R-HRP 10353-046 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol DAB1
Antigen Name Dab, reelin signal transducer, homolog 1 (Drosophila)
Reactivity Human, Mouse, Rat
Conjugation HRP (Horseradish Peroxidase)
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms C630028C02Rik, scrambler, AI956902, scm, yot, scr
Gene ID 1600
Modification phospho-Ser524
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty