|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | CDC25A |
Antigen Name | Cell Division Cycle 25A |
Reactivity | Rat, Mouse, Human |
Conjugation | HRP (Horseradish Peroxidase) |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ImmunoChemistry | Yes |
Antigen Synonyms | M-phase inducer phosphatase 1, CDC25A, Dual specificity phosphatase Cdc25A |
Gene ID | 993 |
Modification | phospho-Ser178 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |