Anti-CDC25A Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3093R-CY5.5
10352-896EA 811.12 USD
10352-896
Anti-CDC25A Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Tyrosine protein phosphatase which functions as a dosage-dependent inducer of mitotic progression. Directly dephosphorylates CDK1 and stimulates its kinase activity. Also dephosphorylates CDK2 in complex with cyclin E, in vitro.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDC25A
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR