|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | CDK1 |
Antigen Name | Cyclin-dependent Kinase 1 |
Reactivity | Human, Mouse, Rat |
Conjugation | Alexa Fluor® 350 |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | CDC2, cell division protein kinase 1, p34 protein kinase, Cell Division Cycle 2, cell cycle controller CDC2, G1 to S and G2 to M, CDC28A, P34CDC2, cell division control protein 2 homolog |
Gene ID | 983 |
Modification | phospho-Thr14 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |