Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CBL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
The proto oncogene c CBL was initially identified as the cellular homologue of v CBL oncogene that induces pre B cell lymphomas and myeloid leukemias in mice. In more recent studies CBL has been shown to be a negative regulator of tyrosine kinase signaling. The ubiquitin ligase activity of CBL leads to the degradation of tyrosine kinases, thus attenuating the signal of receptors. Targets of CBL include activated protein tyrosine kinases belonging to the Src and Syk/Zap 70 families. An additional mechanism to attenuate receptor signaling is thought to be achieved by CBL’s interaction with downstream targets of tyrosine kinases, such as PI 3K and Vav.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CBL
Clonality: Polyclonal
Conjugation: HRP (Horseradish Peroxidase)
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-3089R-HRP 10351-158 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol CBL
Antigen Name Cbl proto-oncogene, E3 Ubiquitin Protein Ligase
Reactivity Mouse, Rat, Human
Conjugation HRP (Horseradish Peroxidase)
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms RNF57, SH3-binding protein CBL-C, CBL-3, RING finger protein 57, Signal transduction protein CBL-C, CBLC
Gene ID 867
Modification phospho-Tyr731
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty