|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | CTNNB1 |
Antigen Name | Catenin beta |
Reactivity | Rat, Human, Mouse |
Conjugation | Cy5® |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | ORF Names: OK/SW-cl.35, Catenin beta, PRO2286, Catenin beta-1, CTNNB, Beta-catenin, CTNNB1 |
Gene ID | 1499 |
Modification | phospho-Ser675 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |