Anti-CAMK2A Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3081R-A555
10350-998EA 776.19 USD
10350-998
Anti-CAMK2A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
CaM-kinase II (CAMK2) is a prominent kinase in the central nervous system that may function in long-term potentiation and neurotransmitter release. Member of the NMDAR signaling complex in excitatory synapses it may regulate NMDAR-dependent potentiation of the AMPAR and synaptic plasticity (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAMK2A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR