|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | CYLD |
Antigen Name | Cylindromatosis 1 (turban tumor syndrome) |
Reactivity | Human, Mouse, Rat |
Conjugation | Cy3® |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | SBS, CDMT, USPL2, CYLDI, MFT1, EAC, TEM, BRSS, CYLD1, MFT |
Gene ID | 1540 |
Modification | phospho-Ser418 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 390-440/956 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |