|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | CSF1R |
Antigen Name | Colony Stimulating Factor 1 receptor |
Reactivity | Rat, Dog, Mouse, Human |
Conjugation | Cy5® |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
ImmunoFluorescence | Yes |
Antigen Synonyms | FIM2, CSF-1R, HDLS, FMS, CSFR, C-FMS, M-CSF-R, CD115 |
Gene ID | 1436 |
Modification | phospho-Tyr723 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 694-744/972 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |