Anti-CD19 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3065R-CY3
10350-740EA 811.12 USD
10350-740
Anti-CD19 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Assembles with the antigen receptor of B-lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD19
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR