Anti-CEBPB Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-3062R-CY5
10350-670EA 746.43 USD
10350-670
Anti-CEBPB Rabbit Polyclonal Antibody (Cy5®)
Antibodies
The protein encoded by this intronless gene is a bZIP transcription factor which can bind as a homodimer to certain DNA regulatory regions. It can also form heterodimers with the related proteins CEBP alpha, CEBP delta, and CEBP gamma. The encoded protein is important in the regulation of genes involved in immune and inflammatory responses and has been shown to bind to the IL1 response element in the IL6 gene, as well as to regulatory regions of several acute-phase and cytokine genes. In addition, the encoded protein can bind the promoter and upstream element and stimulate the expression of the collagen type I gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CEBPB
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR