Anti-CEBPA Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3061R-A647
10350-640EA 811.12 USD
10350-640
Anti-CEBPA Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
C/EBP is a DNA-binding protein that recognizes two different motifs: the CCAAT homology common to many promoters and the enhanced core homology common to many enhancers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CEBPA
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR