Anti-BCL2L11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-3058R-A555
10355-332EA 811.12 USD
10355-332
Anti-BCL2L11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Induces apoptosis and anoikis. The isoforms vary in cytotoxicity with isoform BimS being the most potent and isoform BimEL being the least potent.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL2L11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR