|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | BTK |
Antigen Name | Bruton agammaglobulinemia tyrosine kinase |
Reactivity | Human, Mouse |
Conjugation | FITC (Fluorescein Isothiocyanate) |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | AT, AGMX1, ATK, IMD1, PSCTK1, RP1-164F3.2, BPK, MGC126262, MGC126261, XLA |
Gene ID | 695 |
Modification | phospho-Ser180 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 160-200/659 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |