Anti-TP53BP1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3020R-CY5
10352-700EA 776.17 USD
10352-700
Anti-TP53BP1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Plays a key role in the response to DNA damage. May have a role in checkpoint signaling during mitosis. Enhances TP53-mediated transcriptional activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TP53BP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR