Anti-KLK3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-3003R-A350
10349-858EA 811.12 USD
10349-858
Anti-KLK3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
APS (adapter protein with Pleckstrin homology and Src homology 2 domains) is a member of the Lnk family, an adaptor protein that is involved in B cell signaling, insulin signaling and cytoskeletal reorganisation. A PH and an SH2 domain containing adaptor protein that links activated tyrosine kinases to signaling pathways. It is tyrosine phosphorylated by JAK2, KIT and other kinases during B cell receptor stimulation of many different cytokines, chemokines and leukokines. APS has been shown to inhibit the JAK STAT pathway in collaboration with cCbl.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLK3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR