Anti-FUS Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-2980R-CY3
10355-160EA 776.17 USD
10355-160
Anti-FUS Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Binds both single-stranded and double-stranded DNA and promotes ATP-independent annealing of complementary single-stranded DNAs and D-loop formation in superhelical double-stranded DNA. May play a role in maintenance of genomic integrity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FUS
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR