Anti-F8 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-2974R-CY5
10349-354EA 776.17 USD
10349-354
Anti-F8 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Factor VIII, along with calcium and phospholipid, acts as a cofactor for factor IXa when it converts factor X to the activated form, factor Xa.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Factor VIII
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR