Anti-ABCE1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-2948R-CY7
10348-916EA 775.93 USD
10348-916
Anti-ABCE1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
ABCE1 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the OABP subfamily. It is alternatively referred to as the RNase L inhibitor as it functions to block the activity of ribonuclease L(referenced from entrez gene).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABCE1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR