Anti-FUCA1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-2940R-CY3
10348-766EA 776.17 USD
10348-766
Anti-FUCA1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Alpha-L-fucosidase is responsible for hydrolyzing the alpha-1,6-linked fucose joined to the reducing-end N-acetylglucosamine of the carbohydrate moieties of glycoproteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FUCA1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR