Anti-RASSF2 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-2939R-CY5.5
10354-888EA 811.1 USD
10354-888
Anti-RASSF2 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Potential tumor suppressor. Acts as a KRAS-specific effector protein. May promote apoptosis and cell cycle arrest. Stabilizes STK3/MST2 by protecting it from proteasomal degradation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RASSF2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR