Anti-ARHGEF7 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-2936R-A555
10348-720EA 811.1 USD
10348-720
Anti-ARHGEF7 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
ARHGEF7 is a p21-activated protein kinase (Pak)-interacting exchange factor that has been identified as a putative guanine nucleotide exchange factor (GEF) for Rac/Cdc42. It is expressed ubiquitously in all the tissue examined and other isoforms are expressed mainly in the central nervous system. Regulated phosphorylation of ARHGEF7 is necessary to maintain the balance between normal signalling by EGFR and Src versus aberrant growth and transformation.Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene, but some of their full-length sequences have not been determined.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARHGEF7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR