Anti-TIP47 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-2914R-CY7
10348-350EA 811.1 USD
10348-350
Anti-TIP47 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Required for the transport of mannose 6-phosphate receptors (MPR) from endosomes to the trans-Golgi network.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TIP47
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR