Anti-FOLH1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-2899R-A350
10352-284EA 746.42 USD
10352-284
Anti-FOLH1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Has both folate hydrolase and N-acetylated-alpha-linked-acidic dipeptidase (NAALADase) activity. Has a preference for tri-alpha-glutamate peptides. In the intestine, required for the uptake of folate. In the brain, modulates excitatory neurotransmission through the hydrolysis of the neuropeptide, N-aceylaspartylglutamate (NAAG), thereby releasing glutamate. Isoform PSM-4 and isoform PSM-5 would appear to be physiologically irrelevant. Involved in prostate tumor progression. Also exhibits a dipeptidyl-peptidase IV type activity. In vitro, cleaves Gly-Pro-AMC.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PMSA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR